Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ine | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057665.5 | NP_477013.1 | 1 - 2 | 242 | 69 | |
GenBank mRNA | BT099793.1 | - | 1 - 2 | 225 | 69 | |
U66460.1 | - | 1 - 2 | 201 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |