Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057686.4 | NP_477034.1 | 3 - 4 | 432 | 83 | |
GenBank mRNA | BT089017.1 | - | 3 - 4 | 384 | 83 | |
KX531942.1 | - | 2 - 3 | 295 | 83 | ||
U08217.1 | - | 3 - 4 | 410 | 83 | ||
U24490.1 | - | 3 - 4 | 407 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |