Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103908.2 | NP_001097378.1 | 11 - 12 | 1837 | 118 | |
NM_001299684.1 | NP_001286613.1 | 12 - 13 | 1882 | 118 | ||
NM_057699.5 | NP_477047.1 | 11 - 12 | 1745 | 118 | ||
GenBank mRNA | AY051678.1 | - | 10 - 11 | 1721 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |