Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103908.2 | NP_001097378.1 | 3 - 4 | 489 | 79 | |
NM_001299684.1 | NP_001286613.1 | 4 - 5 | 534 | 79 | ||
NM_057699.5 | NP_477047.1 | 3 - 4 | 397 | 79 | ||
NM_057700.4 | NP_477048.1 | 3 - 4 | 489 | 79 | ||
GenBank mRNA | AY051678.1 | - | 2 - 3 | 373 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |