Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103908.2 | NP_001097378.1 | 5 - 6 | 688 | 82 | |
NM_001299684.1 | NP_001286613.1 | 6 - 7 | 733 | 82 | ||
NM_057699.5 | NP_477047.1 | 5 - 6 | 596 | 82 | ||
NM_057700.4 | NP_477048.1 | 5 - 6 | 688 | 82 | ||
GenBank mRNA | AY051678.1 | - | 4 - 5 | 572 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |