Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057716.4 | NP_477064.1 | 1 | 2366 | 139 | |
NM_165597.2 | NP_724659.1 | 1 | 2948 | 139 | ||
GenBank mRNA | AY058663.1 | - | 1 | 2319 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |