Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057732.4 | NP_477080.1 | 1 - 2 | 284 | 61 | |
GenBank mRNA | AY061049.1 | - | 1 - 2 | 241 | 61 | |
X99511.1 | - | 260 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |