Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273836.1 | NP_001260765.1 | 2 - 3 | 1495 | 90 | |
NM_057744.3 | NP_477092.1 | 2 - 3 | 1495 | 90 | ||
GenBank mRNA | BT044167.1 | - | 2 - 3 | 1474 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |