Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssb-c31a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057788.4 | NP_477136.1 | 1 - 2 | 159 | 72 | |
GenBank mRNA | BT023261.1 | - | 1 - 2 | 131 | 72 | |
KX531387.1 | - | 1 - 2 | 60 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |