Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: toc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042864.2 | NP_001036329.1 | 6 - 7 | 1527 | 68 | |
NM_001273058.1 | NP_001259987.1 | 3 - 4 | 1005 | 68 | ||
NM_001273059.1 | NP_001259988.1 | 6 - 7 | 1527 | 68 | ||
NM_001273060.1 | NP_001259989.1 | 5 - 6 | 1057 | 68 | ||
NM_001273061.1 | NP_001259990.1 | 5 - 6 | 1368 | 68 | ||
NM_057805.3 | NP_477153.1 | 8 - 9 | 5037 | 68 | ||
NM_164524.2 | NP_722875.2 | 6 - 7 | 1201 | 68 | ||
NM_164525.2 | NP_722876.1 | 4 - 5 | 912 | 68 | ||
NM_164526.2 | NP_722877.1 | 3 - 4 | 1594 | 68 | ||
GenBank mRNA | BT031283.1 | - | 6 - 7 | 1185 | 68 | |
BT099598.1 | - | 4 - 5 | 912 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |