Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273199.1 | NP_001260128.1 | 3 - 4 | 667 | 78 | |
NM_001273200.1 | NP_001260129.1 | 3 - 4 | 765 | 78 | ||
NM_057810.4 | NP_477158.1 | 3 - 4 | 825 | 78 | ||
GenBank mRNA | AY052000.1 | - | 3 - 4 | 805 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |