Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273234.1 | NP_001260163.1 | 3 - 4 | 1099 | 121 | |
NM_057819.4 | NP_477167.1 | 3 - 4 | 1099 | 121 | ||
GenBank mRNA | BT044185.1 | - | 3 - 4 | 1069 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |