Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vang | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273890.2 | NP_001260819.1 | 1 - 2 | 575 | 81 | |
NM_057829.4 | NP_477177.1 | 1 - 2 | 575 | 81 | ||
GenBank mRNA | BT150320.1 | - | 1 - 2 | 569 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |