Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299231.1 | NP_001286160.1 | 5 - 6 | 2424 | 66 | |
NM_057837.5 | NP_477185.1 | 5 - 6 | 2156 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |