Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057884.5 | NP_477232.1 | 2 - 3 | 575 | 70 | |
GenBank mRNA | AF043263.1 | - | 2 - 3 | 537 | 70 | |
BT044099.1 | - | 2 - 3 | 574 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |