Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057920.4 | NP_477268.2 | 6 - 7 | 2251 | 90 | |
GenBank mRNA | AY061410.1 | - | 6 - 7 | 2232 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |