Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169159.2 | NP_001162630.1 | 5 - 6 | 2221 | 90 | |
NM_001169160.2 | NP_001162631.1 | 6 - 7 | 1589 | 90 | ||
NM_001297828.1 | NP_001284757.1 | 6 - 7 | 1723 | 90 | ||
NM_001297829.1 | NP_001284758.1 | 6 - 7 | 1723 | 90 | ||
NM_057941.5 | NP_477289.5 | 6 - 7 | 1658 | 90 | ||
NM_057942.5 | NP_477290.6 | 6 - 7 | 1775 | 90 | ||
NM_134314.4 | NP_599141.5 | 6 - 7 | 1723 | 90 | ||
NM_134315.4 | NP_599142.5 | 6 - 7 | 1775 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |