Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297881.1 | NP_001284810.1 | 2 - 3 | 554 | 66 | |
NM_057986.4 | NP_477334.2 | 2 - 3 | 771 | 66 | ||
GenBank mRNA | BT010002.1 | - | 2 - 3 | 769 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |