Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258605.1 | NP_001245534.1 | 4 - 5 | 583 | 65 | |
NM_001297972.1 | NP_001284901.1 | 4 - 5 | 718 | 65 | ||
NM_080053.4 | NP_524792.1 | 4 - 5 | 718 | 65 | ||
GenBank mRNA | AB042625.2 | - | 4 - 5 | 636 | 65 | |
AY070980.1 | - | 4 - 5 | 716 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |