Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258605.1 | NP_001245534.1 | 6 - 7 | 1177 | 61 | |
NM_080053.4 | NP_524792.1 | 6 - 7 | 1312 | 61 | ||
GenBank mRNA | AB042625.2 | - | 6 - 7 | 1230 | 61 | |
AY070980.1 | - | 6 - 7 | 1310 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Drosophila melanogaster |