Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080071.3 | NP_524810.2 | 1 - 2 | 599 | 62 | |
GenBank mRNA | BT029035.1 | - | 1 - 2 | 579 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |