Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103847.2 | NP_001097317.1 | 11 - 12 | 1979 | 61 | |
NM_001274051.1 | NP_001260980.1 | 10 - 11 | 1567 | 61 | ||
NM_080074.4 | NP_524813.2 | 10 - 11 | 2100 | 61 | ||
NM_137146.3 | NP_610990.2 | 10 - 11 | 2097 | 61 | ||
GenBank mRNA | BT150395.1 | - | 10 - 11 | 2098 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |