Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103847.2 | NP_001097317.1 | 7 - 8 | 1085 | 143 | |
NM_080074.4 | NP_524813.2 | 6 - 7 | 1206 | 143 | ||
NM_137146.3 | NP_610990.2 | 6 - 7 | 1206 | 143 | ||
NM_166070.3 | NP_725419.2 | 6 - 7 | 1206 | 143 | ||
GenBank mRNA | BT150395.1 | - | 6 - 7 | 1204 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |