Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274419.2 | NP_001261348.1 | 1 - 2 | 111 | 98 | |
NM_080118.2 | NP_524857.3 | 1 - 2 | 200 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |