Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274419.2 | NP_001261348.1 | 2 - 3 | 386 | 75 | |
NM_080118.2 | NP_524857.3 | 2 - 3 | 475 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |