Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043076.3 | NP_001036541.1 | 2 - 3 | 429 | 72 | |
NM_001043080.3 | NP_001036545.1 | 2 - 3 | 359 | 72 | ||
NM_080153.5 | NP_524892.2 | 2 - 3 | 551 | 72 | ||
GenBank mRNA | BT056247.1 | - | 2 - 3 | 528 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |