Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043076.3 | NP_001036541.1 | 4 - 5 | 598 | 56 | |
NM_001043077.3 | NP_001036542.1 | 2 - 3 | 237 | 56 | ||
NM_001043078.2 | NP_001036543.1 | 2 - 3 | 275 | 56 | ||
NM_001043079.2 | NP_001036544.1 | 2 - 3 | 275 | 56 | ||
NM_001043080.3 | NP_001036545.1 | 4 - 5 | 528 | 56 | ||
NM_001202010.4 | NP_001188939.1 | 4 - 5 | 1766 | 56 | ||
NM_001202012.2 | NP_001188941.1 | 2 - 3 | 275 | 56 | ||
NM_001274055.1 | NP_001260984.1 | 4 - 5 | 1787 | 56 | ||
NM_001274056.2 | NP_001260985.1 | 2 - 3 | 136 | 56 | ||
NM_001274057.2 | NP_001260986.1 | 2 - 3 | 237 | 56 | ||
NM_001274058.2 | NP_001260987.1 | 2 - 3 | 237 | 56 | ||
NM_080153.5 | NP_524892.2 | 4 - 5 | 720 | 56 | ||
GenBank mRNA | BT056247.1 | - | 4 - 5 | 697 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |