Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043076.3 | NP_001036541.1 | 5 - 6 | 687 | 83 | |
NM_001043077.3 | NP_001036542.1 | 3 - 4 | 326 | 83 | ||
NM_001043078.2 | NP_001036543.1 | 3 - 4 | 364 | 83 | ||
NM_001043079.2 | NP_001036544.1 | 3 - 4 | 364 | 83 | ||
NM_001043080.3 | NP_001036545.1 | 5 - 6 | 617 | 83 | ||
NM_001202010.4 | NP_001188939.1 | 5 - 6 | 1855 | 83 | ||
NM_001202012.2 | NP_001188941.1 | 3 - 4 | 364 | 83 | ||
NM_001274055.1 | NP_001260984.1 | 5 - 6 | 1876 | 83 | ||
NM_001274056.2 | NP_001260985.1 | 3 - 4 | 225 | 83 | ||
NM_001274057.2 | NP_001260986.1 | 3 - 4 | 326 | 83 | ||
NM_001274058.2 | NP_001260987.1 | 3 - 4 | 326 | 83 | ||
NM_080153.5 | NP_524892.2 | 5 - 6 | 809 | 83 | ||
GenBank mRNA | BT056247.1 | - | 5 - 6 | 786 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |