Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043076.3 | NP_001036541.1 | 7 - 8 | 1839 | 65 | |
NM_001043077.3 | NP_001036542.1 | 5 - 6 | 1478 | 65 | ||
NM_001043078.2 | NP_001036543.1 | 5 - 6 | 1516 | 65 | ||
NM_001202010.4 | NP_001188939.1 | 7 - 8 | 3007 | 65 | ||
NM_001274055.1 | NP_001260984.1 | 7 - 8 | 3028 | 65 | ||
NM_001274056.2 | NP_001260985.1 | 5 - 6 | 1377 | 65 | ||
NM_001274057.2 | NP_001260986.1 | 5 - 6 | 1478 | 65 | ||
NM_001274058.2 | NP_001260987.1 | 5 - 6 | 1478 | 65 | ||
NM_080153.5 | NP_524892.2 | 7 - 8 | 1961 | 65 | ||
GenBank mRNA | BT056247.1 | - | 7 - 8 | 1938 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |