Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298855.1 | NP_001285784.1 | 5 - 6 | 1086 | 90 | |
NM_080156.4 | NP_524895.2 | 5 - 6 | 993 | 90 | ||
GenBank mRNA | AY058729.1 | - | 5 - 6 | 982 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |