Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298855.1 | NP_001285784.1 | 6 - 7 | 1208 | 74 | |
NM_080156.4 | NP_524895.2 | 6 - 7 | 1115 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |