Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080262.4 | NP_525001.2 | 2 - 3 | 412 | 67 | |
GenBank mRNA | AJ249345.1 | - | 2 - 3 | 249 | 67 | |
AY051708.1 | - | 2 - 3 | 378 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |