Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mei-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297930.1 | NP_001284859.1 | 3 - 4 | 1036 | 102 | |
NM_080329.3 | NP_525068.1 | 4 - 5 | 1151 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |