Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169239.2 | NP_001162710.2 | 9 - 10 | 4114 | 78 | |
NM_080341.4 | NP_525080.3 | 9 - 10 | 4114 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |