Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samuel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273443.3 | NP_001260372.1 | 2 - 3 | 1215 | 60 | |
NM_001273444.3 | NP_001260373.2 | 2 - 3 | 1215 | 60 | ||
NM_001298899.1 | NP_001285828.1 | 2 - 3 | 616 | 60 | ||
NM_080362.4 | NP_525101.2 | 2 - 3 | 616 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |