Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NaPi-T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274052.2 | NP_001260981.1 | 3 - 4 | 570 | 72 | |
NM_080377.3 | NP_525116.2 | 3 - 4 | 904 | 72 | ||
GenBank mRNA | AY071092.1 | - | 3 - 4 | 567 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |