Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mRpS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080380.3 | NP_525119.1 | 1 - 2 | 185 | 67 | |
GenBank mRNA | AY084147.1 | - | 1 - 2 | 179 | 67 | |
KX532008.1 | - | 1 - 2 | 111 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |