Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259641.1 | NP_001246570.1 | 11 - 12 | 5029 | 89 | |
NM_001259642.1 | NP_001246571.1 | 11 - 12 | 5029 | 89 | ||
NM_001259643.1 | NP_001246572.1 | 11 - 12 | 5038 | 89 | ||
NM_001259644.1 | NP_001246573.1 | 11 - 12 | 5029 | 89 | ||
NM_001259645.1 | NP_001246574.1 | 11 - 12 | 5029 | 89 | ||
NM_001259646.1 | NP_001246575.1 | 10 - 11 | 5008 | 89 | ||
NM_001259647.1 | NP_001246576.1 | 10 - 11 | 4437 | 89 | ||
NM_001259649.1 | NP_001246578.1 | 11 - 12 | 5029 | 89 | ||
NM_001299992.1 | NP_001286921.1 | 11 - 12 | 5029 | 89 | ||
GenBank mRNA | BT133165.1 | - | 11 - 12 | 5034 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |