Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297973.1 | NP_001284902.1 | 3 - 4 | 912 | 112 | |
NM_001297974.1 | NP_001284903.1 | 3 - 4 | 1346 | 112 | ||
NM_080525.3 | NP_536786.1 | 4 - 5 | 1186 | 112 | ||
GenBank mRNA | AY122270.1 | - | 4 - 5 | 1190 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |