Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201735.2 | NP_001188664.1 | 4 - 5 | 1280 | 66 | |
NM_001298463.1 | NP_001285392.1 | 3 - 4 | 1230 | 66 | ||
NM_133018.4 | NP_573246.1 | 4 - 5 | 1340 | 66 | ||
GenBank mRNA | BT023820.1 | - | 4 - 5 | 1302 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |