Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038764.2 | NP_001033853.1 | 4 - 5 | 721 | 90 | |
NM_001272744.1 | NP_001259673.1 | 4 - 5 | 583 | 90 | ||
NM_001272745.1 | NP_001259674.1 | 4 - 5 | 579 | 90 | ||
NM_133043.3 | NP_573271.1 | 4 - 5 | 675 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |