Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: upd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370039.1 | NP_001356882.1 | 2 - 3 | 614 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |