Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175975.2 | NP_787989.1 | 3 - 4 | 440 | 82 | |
NM_175976.2 | NP_787990.1 | 3 - 4 | 479 | 82 | ||
NM_175977.2 | NP_787991.1 | 3 - 4 | 723 | 82 | ||
NM_175978.2 | NP_787992.1 | 2 - 3 | 1210 | 82 | ||
GenBank mRNA | AY060782.1 | - | 2 - 3 | 1189 | 82 | |
BT133408.1 | - | 3 - 4 | 479 | 82 | ||
L33475.1 | - | 3 - 4 | 394 | 82 | ||
L33784.1 | - | 2 - 3 | 609 | 82 | ||
U11442.2 | - | 2 - 3 | 1228 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |