Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTPase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058022.5 | NP_477370.1 | 2 - 3 | 347 | 73 | |
NM_164513.4 | NP_722860.1 | 2 - 3 | 362 | 73 | ||
NM_164514.3 | NP_722861.1 | 1 - 2 | 433 | 73 | ||
NM_164515.3 | NP_722862.1 | 2 - 3 | 358 | 73 | ||
GenBank mRNA | AY061134.1 | - | 2 - 3 | 334 | 73 | |
BT133416.1 | - | 2 - 3 | 356 | 73 | ||
EU852293.1 | - | 2 - 3 | 161 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |