Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrapper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058056.4 | NP_477404.1 | 4 - 5 | 1472 | 77 | |
GenBank mRNA | AY047524.1 | - | 4 - 5 | 1597 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |