Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169462.2 | NP_001162933.1 | 5 - 6 | 1512 | 104 | |
NM_058074.4 | NP_477422.2 | 6 - 7 | 1355 | 104 | ||
NM_058075.4 | NP_477423.1 | 6 - 7 | 1243 | 104 | ||
NM_058076.4 | NP_477424.1 | 4 - 5 | 1013 | 104 | ||
NM_134320.3 | NP_599147.1 | 6 - 7 | 1243 | 104 | ||
NM_134321.3 | NP_599148.1 | 4 - 5 | 1013 | 104 | ||
NM_134322.3 | NP_599149.1 | 4 - 5 | 1013 | 104 | ||
GenBank mRNA | AF144232.1 | - | 5 - 6 | 786 | 104 | |
AF161257.1 | - | 4 - 5 | 1024 | 104 | ||
AF161259.1 | - | 4 - 5 | 1024 | 104 | ||
BT025053.1 | - | 5 - 6 | 1441 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |