Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169609.2 | NP_001163080.1 | 1 - 2 | 608 | 66 | |
NM_058096.4 | NP_477444.1 | 3 - 4 | 845 | 66 | ||
GenBank mRNA | BT004883.1 | - | 1 - 2 | 591 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |