Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058135.4 | NP_477483.1 | 1 | 3503 | 98 | |
GenBank mRNA | AY071147.1 | - | 1 | 3501 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |