Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058151.5 | NP_477499.1 | 1 - 2 | 195 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |