Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299643.1 | NP_001286572.1 | 2 - 3 | 983 | 67 | |
NM_133166.4 | NP_573394.1 | 1 - 2 | 785 | 67 | ||
GenBank mRNA | AY051558.1 | - | 1 - 2 | 736 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |